missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CD112 / Nectin-2 / PVRL2 Protein

A DNA sequence encoding the Homo sapiens (Human) CD112 / Nectin-2 / PVRL2, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7863-ec-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16011251

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_001036189.1
CD112 / Nectin-2 / PVRL2 Protein
Research Use Only
PVRL2
Recombinant Protein
E. coli
QDVRVQVLPE VRGQLGGTVE LPCHLLPPVP GLYISLVTWQ RPDAPANHQN VAAFHPKMGP SFPSPKPGSE RLSFVSAKQS TGQDTEAELQ DATLALHGLT VEDEGNYTCE FATFPKGSVR GMTWLRVIAK PKNQAEAQKV TFSQDPTTVA LCISKEGRPP ARISWLSSLD WEAKETQVSG TLAGTVTVTS RFTLVPSGRA DGVTVTCKVE HESFEEPALI PVTLSVRYPP EVSISGYDDN WYLGRTDATL SCDVRSNPEP TGYDWSTTSG TFPTSAVAQG SQLVIHAVDS LFNTTFVCTV TNAVGMGRAE QVIFVRETPN TAGAGATGG
Greater than 90% as determined by SDS-PAGE.
5819
10 μg
Not determined.
Extracellular Domain
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers