missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CCL3 / Mip1a Protein
Specifications
Specifications
| Accession Number | NP_002974.1 |
| Gene ID (Entrez) | 6348 |
| Name | CCL3 / Mip1a Protein |
| Quantity | 5 μg |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | Less than 1.0 EU/ug as determined by LAL method. |
| Gene Symbol | CCL3 |
| Protein Length | Partial (see sequence information for more details). |
| Biological Activity | The biological activity determined by a chemotaxis bioassay using human monocytes is in a concentration range of 1.0-10 ng/ml. |
| Product Type | Recombinant Protein |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?