missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CASP7 / caspase 7 / MCH3 Protein
A DNA sequence encoding the Homo sapiens (Human) CASP7 / caspase 7 / MCH3, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8834-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001218.1 | |
CASP7 / caspase 7 / MCH3 Protein | |
Research Use Only | |
CASP7 | |
Recombinant Protein | |
E. coli | |
ANPRYKIPVE ADFLFAYSTV PGYYSWRSPG RGSWFVQALC SILEEHGKDL EIMQILTRVN DRVARHFESQ SDDPHFHEKK QIPCVVSMLT KELYFSQ | |
Greater than 90% as determined by SDS-PAGE. |
840 | |
50 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |