missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CASP7 / caspase 7 / MCH3 Protein

A DNA sequence encoding the Homo sapiens (Human) CASP7 / caspase 7 / MCH3, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8834-ec-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16036731

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_001218.1
CASP7 / caspase 7 / MCH3 Protein
Research Use Only
CASP7
Recombinant Protein
E. coli
ANPRYKIPVE ADFLFAYSTV PGYYSWRSPG RGSWFVQALC SILEEHGKDL EIMQILTRVN DRVARHFESQ SDDPHFHEKK QIPCVVSMLT KELYFSQ
Greater than 90% as determined by SDS-PAGE.
840
500 μg
Not determined.
Partial (see sequence information for more details).
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers