Learn More
enQuireBio™ Recombinant Human Calcineurin B homologous protein 1
A DNA sequence encoding the Homo sapiens (Human) Calcineurin B homologous protein 3, was expressed in the hosts and tags indicated.
Brand:  enQuireBio™ QP6772-ec-1mg
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 16015730
  • 24050.00 NOK / 1mg
Estimated Shipment: 09-07-2024
Specifications
NP_001161797.1
Calcineurin B homologous protein 1
Research Use Only
TESC
Recombinant Protein
E. coli
GAAHSASEEV RELEGKTGFS SDQIEQLHRR FKQLSGDQPT IRKENFNNVP DLELNPIRSK IVRAFFDNRN LRKGPSGLAD EINFEDFLTI MSYFRPIDTT MDEEQVELSR KEKLRFLFHM YDSDSDGRIT LEEYRNVVEE LLSGNPHIEK ESARSIADGA MMEAASVCMG QMEPDQVYEG ITFEDFLKIW QGIDIETKMH VRFLNMETMA LCH
Greater than 90% as determined by SDS-PAGE.
54997
1 mg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol