missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Calcineurin B homologous protein 1
A DNA sequence encoding the Homo sapiens (Human) Calcineurin B homologous protein 3, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6772-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001161797.1 | |
Calcineurin B homologous protein 1 | |
Research Use Only | |
TESC | |
Recombinant Protein | |
E. coli | |
GAAHSASEEV RELEGKTGFS SDQIEQLHRR FKQLSGDQPT IRKENFNNVP DLELNPIRSK IVRAFFDNRN LRKGPSGLAD EINFEDFLTI MSYFRPIDTT MDEEQVELSR KEKLRFLFHM YDSDSDGRIT LEEYRNVVEE LLSGNPHIEK ESARSIADGA MMEAASVCMG QMEPDQVYEG ITFEDFLKIW QGIDIETKMH VRFLNMETMA LCH | |
Greater than 90% as determined by SDS-PAGE. |
54997 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |