missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human C1QTNF2 Protein
A cDNA sequence encoding the C1QTNF2 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11231-1mg
Additional Details : Weight : 0.01000kg
Specifications
C1QTNF2 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MKHHHHHHAS DPLLGAFARR DFRKGSPQLV CSLPGPQGPP GPPGAPGPSG MMGRMGFPGK DGQDGHDGDR GDSGEEGPPG DPLLGAFARR DFRKGSPQLV CSLPGPQGPP GPPGAPGPSG MMGRMGFPGK DGQDGHDGDR GDSGEEGPPG RTGNRGKPGP KGKAGAIGRA GPRGPKGVNG TPGKHGTPGK KGPKGKKGEP GLPGPCSCGS GHTKSAFSVA VTKSYPRERL PIKFDKILMN EGGHYNASSG KFVCGVPGIY YFTYDITLAN KHLAIGLVHN GQYRIRTFDA NTGNHDVASG STILALKQGD EVWLQIFYSE QNGLFYDPYW TDSLFTGFLI YADQDDPNEV |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
C1QTNF2 Human was filtered (0.4microm) and lyophilized from 0.5 mg/ml supplied in 0.03M Acetate buffer, pH 4. |