missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Breast cancer metastasis-suppressor 1 Protein
A DNA sequence encoding the Homo sapiens (Human) Breast cancer metastasis-suppressor 1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8482-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_056214.1 | |
Breast cancer metastasis-suppressor 1 Protein | |
Research Use Only | |
BRMS1 | |
Recombinant Protein | |
E. coli | |
MPVQPPSKDT EEMEAEGDSA AEMNGEEEES EEERSGSQTE SEEESSEMDD EDYERRRSEC VSEMLDLEKQ FSELKEKLFR ERLSQLRLRL EEVGAERAPE YTEPLGGLQR SLKIRIQVAG IYKGFCLDVI RNKYECELQG AKQHLESEKL LLYDTLQGEL QERIQRLEED RQSLDLSSEW WDDKLHARGS SRSWDSLPPS KRKKAPLVSG PYIVYMLQEI DILEDWTAIK KARAA | |
Greater than 90% as determined by SDS-PAGE. |
25855 | |
50 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |