Learn More
enQuireBio™ Recombinant Human Breast cancer metastasis-suppressor 1 Protein
A DNA sequence encoding the Homo sapiens (Human) Breast cancer metastasis-suppressor 1, was expressed in the hosts and tags indicated.
Brand:  enQuireBio™ QP8482-ec-100ug
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 16024861
  • 5325.00 NOK / 100µg
Estimated Shipment: 08-07-2024
Specifications
NP_056214.1
Breast cancer metastasis-suppressor 1 Protein
Research Use Only
BRMS1
Recombinant Protein
E. coli
MPVQPPSKDT EEMEAEGDSA AEMNGEEEES EEERSGSQTE SEEESSEMDD EDYERRRSEC VSEMLDLEKQ FSELKEKLFR ERLSQLRLRL EEVGAERAPE YTEPLGGLQR SLKIRIQVAG IYKGFCLDVI RNKYECELQG AKQHLESEKL LLYDTLQGEL QERIQRLEED RQSLDLSSEW WDDKLHARGS SRSWDSLPPS KRKKAPLVSG PYIVYMLQEI DILEDWTAIK KARAA
Greater than 90% as determined by SDS-PAGE.
25855
100 μg
Not determined.
Partial (see sequence information for more details).
Human
GST
Tris-based buffer, 50% glycerol