missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human BCL-W / BCL2L2 Protein
A DNA sequence encoding the Homo sapiens (Human) BCL-W / BCL2L2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7839-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001186768.1 | |
BCL-W / BCL2L2 Protein | |
Research Use Only | |
BCL2L2 | |
Recombinant Protein | |
E. coli | |
ATPASAPDTR ALVADFVGYK LRQKGYVCGA GPGEGPAADP LHQAMRAAGD EFETRFRRTF SDLAAQLHVT PGSAQQRFTQ VSDELFQGGP NWGRLVAFFV FGAALCAESV NKEMEPLVGQ VQEWMVAYLE TQLADWIHSS GGWAEFTALY GDGALEEARR LREGNWASVR TVLTGAVALG ALVTVGAFFA SK | |
Greater than 90% as determined by SDS-PAGE. |
599 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |