missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human B-cell receptor-associated protein 31 Protein
A DNA sequence encoding the Homo sapiens (Human) B-cell receptor-associated protein 31, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8444-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001132913.1 | |
B-cell receptor-associated protein 31 Protein | |
Research Use Only | |
BCAP31 | |
Recombinant Protein | |
E. coli | |
SLQWTAVATF LYAEVFVVLL LCIPFISPKR WQKIFKSRLV ELLVSYGNTF FVVLIVILVL LVIDAVREIR KYDDVTEKVN LQNNPGAMEH FHMKLFRAQR NLYIAGFSLL LSFLLRRLVT LISQQATLLA SNEAFKKQAE SASEAAKKYM EENDQLKKGA AVDGGKLDVG NAEVKLEEEN RSLKADLQKL KDELASTKQK LEKAENQVLA MRKQSEGLTK EYDRLLEEHA KLQAAVDGPM DK | |
Greater than 80% as determined by SDS-PAGE. |
10134 | |
200 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |