missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ATP-dependent RNA helicase DDX19A Protein
A DNA sequence encoding the Homo sapiens (Human) ATP-dependent RNA helicase DDX19A, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8225-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001307451.1 | |
ATP-dependent RNA helicase DDX19A Protein | |
Research Use Only | |
DDX19A | |
Recombinant Protein | |
E. coli | |
MATDSWALAV DEQEAAVKSM TNLQIKEEKV KADTNGIIKT STTAEKTDEE EKEDRAAQSL LNKLIRSNLV DNTNQVEVLQ RDPNSPLYSV KSFEELRLKP QLLQGVYAMG FNRPSKIQEN ALPMMLAEPP QNLIAQSQSG TGKTAAFVLA MLSRVEPSDR YPQCLCLSPT YELALQTGKV IEQMGKFYPE LKLAYAVRGN KLERGQKISE QIVIGTPGTV LDWCSKLKFI DPKKIKVFVL DEADVMIATQ GHQDQSIRIQ RMLPRNCQML LFSATFEDSV WKFAQKVVPD PNVIKLKREE ETLDTIKQYY VLCSSRDEKF QALCNLYGAI TIAQAMIFCH TRKTASWLAA ELSKEGHQVA LLSGEMMVEQ RAAVIERFRE GKEKVLVTTN VCARGIDVEQ VSVVINFDLP VDKDGNPDNE TYLHRIGRTG RFGKRGLAVN MVDSKHSMNI LNRIQEHFNK KIERLDTDDL DEIEKIAN | |
Greater than 90% as determined by SDS-PAGE. |
55308 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |