missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ATG14 Protein
A DNA sequence encoding the Homo sapiens (Human) ATG14, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7740-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_055739.2 | |
ATG14 Protein | |
Research Use Only | |
ATG14 | |
Recombinant Protein | |
E. coli | |
MASPSGKGAR ALEAPGCGPR PLARDLVDSV DDAEGLYVAV ERCPLCNTTR RRLTCAKCVQ SGDFVYFDGR DRERFIDKKE RLSRLKSKQE EFQKEVLKAM EGKWITDQLR WKIMSCKMRI EQLKQTICKG NEEMEKNSEG LLKTKEKNQK LYSRAQRHQE KKEKIQRHNR KLGDLVEKKT IDLRSHYERL ANLRRSHILE LTSVIFPIEE VKTGVRDPAD VSSESDSAMT SSTVSKLAEA RRTTYLSGRW VCDDHNGDTS ISITGPWISL PNNGDYSAYY SWVEEKKTTQ GPDMEQSNPA YTISAALCYA TQLVNILSHI LDVNLPKKLC NSEFCGENLS KQKFTRAVKK LNANILYLCF SQHVNLDQLQ PLHTLRNLMY LVSPSSEHLG RSGPFEVRAD LEESMEFVDP GVAGESDESG DERVSDEETD LGTDWENLPS PRFCDIPSQS VEVSQSQSTQ ASPPIASSSA GGMISSAAAS VTSWFKAYTG HR | |
Greater than 90% as determined by SDS-PAGE. |
22863 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |