missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Annexin A1 Protein

A DNA sequence encoding the Homo sapiens (Human) Annexin A1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8765-ec-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16076371

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_000691.1
Annexin A1 Protein
Research Use Only
ANX1
Recombinant Protein
E. coli
SEFLKQAWFI ENEEQEYVQT VKSSKGGPGS AVSPYPTFNP SSDVAALHKA IMVKGVDEAT IIDILTKRNN AQRQQIKAAY LQETGKPLDE TLKKALTGHL EEVVLALLKT PAQFDADELR AAMKGLGTDE DTLIEILASR TNKEIRDINR VYREELKRDL AKDITSDTSG DFRNALLSLA KGDRSEDFGV NEDLADSDAR ALYEAGERRK GTDVNVFNTI LTTRSYPQLR RVFQKYTKYS KHDMNKVLDL ELKGDIEKCL TAIVKCATSK PAFFAEKLHQ AMKGVGTRHK ALIRIMVSRS EIDMNDIKAF YQKMYGISLC QAILDETKGD YEKILVALCG GN
Greater than 90% as determined by SDS-PAGE.
301
50 μg
Not determined.
Partial (see sequence information for more details).
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers