missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Angiogenin Protein
A DNA sequence encoding the Homo sapiens (Human) Angiogenin, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8511-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001091046.1 | |
Angiogenin Protein | |
Research Use Only | |
RNASE5 | |
Recombinant Protein | |
E. coli | |
DNSRYTHFLT QHYDAKPQGR DDRYCESIMR RRGLTSPCKD INTFIHGNKR SIKAICENKN GNPHRENLRI SKSSFQVTTC KLHGGSPWPP CQYRATAGFR NVVVACENGL PVHLDQSIFR RP | |
Greater than 80% as determined by SDS-PAGE. |
283 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |