missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Anaphase-promoting complex subunit 5 Protein
A DNA sequence encoding the Homo sapiens (Human) Anaphase-promoting complex subunit 5, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8491-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001131031.1 | |
Anaphase-promoting complex subunit 5 Protein | |
Research Use Only | |
ANAPC5 | |
Recombinant Protein | |
E. coli | |
ASVHESLYFN PMMTNGVVHA NVFGIKDWVT PYKIAVLVLL NEMSRTGEGA VSLMERRRLN QLLLPLLQGP DITLSKLYKL IEESCPQLAN SVQIRIKLMA EGELKDMEQF FDDLSDSFSG TEPEVHKTSV VGLFLRHMIL AYSKLSFSQV FKLYTALQQY FQNGEKKTVE DADMELTSRD EGERKMEKEE LDVSVREEEV SCSGPLSQKQ AEFFLSQQAS LLKNDETKAL T | |
Greater than 90% as determined by SDS-PAGE. |
51433 | |
200 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |