missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Anaphase-promoting complex subunit 10 Protein
A DNA sequence encoding the Homo sapiens (Human) Anaphase-promoting complex subunit 10, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8254-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001243635.1 | |
Anaphase-promoting complex subunit 10 Protein | |
Research Use Only | |
ANAPC10 | |
Recombinant Protein | |
E. coli | |
TTPNKTPPGA DPKQLERTGT VREIGSQAVW SLSSCKPGFG VDQLRDDNLE TYWQSDGSQP HLVNIQFRRK TTVKTLCIYA DYKSDESYTP SKISVRVGNN FHNLQEIRQL ELVEPSGWIH VPLTDNHKKP TRTFMIQIAV LANHQNGRDT HMRQIKIYTP VEESSIGKFP RCTTIDFMMY RSIR | |
Greater than 80% as determined by SDS-PAGE. |
10393 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |