missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Ameloblastin Protein

A DNA sequence encoding the Homo sapiens (Human) Ameloblastin, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP9909-ye-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16038971

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_057603.1
Ameloblastin Protein
Research Use Only
AMBN
Recombinant Protein
Yeast
VPFFPQQSGT PGMASLSLET MRQLGSLQRL NTLSQYSRYG FGKSFNSLWM HGLLPPHSSL PWMRPREHET QQYEYSLPVH PPPLPSQPSL KPQQPGLKPF LQSAAATTNQ ATALKEALQP PIHLGHLPLQ EGELPLVQQQ VAPSDKPPKP ELPGVDFADP QGPSLPGMDF PDPQGPSLPG LDFADPQGST IFQIARLISH GPMPQNKQSP LYPGMLYVPF GANQLNAPAR LGIMSSEEVA GGREDPMAYG AMFPGFGGMR PGFEGMPHNP AMGGDFTLEF DSPVAATKGP ENEEGGAQGS PMPEANPDNL ENPAFLTELE PAPHAGLLAL PKDDIPGLPR SPSGKMKGLP SVTPAAADPL MTPELADVYR TYDADMTTSV DFQEEATMDT TMAPNSLQTS MPGNKAQEPE MMHDAWHFQE P
Greater than 90% as determined by SDS-PAGE.
258
10 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers