missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human ALDH1A3 / RALDH3 Protein

A DNA sequence encoding the Homo sapiens (Human) ALDH1A3 / RALDH3, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP5644-ye-200ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15936679

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_000684.2
ALDH1A3 / RALDH3 Protein
Research Use Only
ALDH1A3
Recombinant Protein
Yeast
ATANGAVENG QPDRKPPALP RPIRNLEVKF TKIFINNEWH ESKSGKKFAT CNPSTREQIC EVEEGDKPDV DKAVEAAQVA FQRGSPWRRL DALSRGRLLH QLADLVERDR ATLAALETMD TGKPFLHAFF IDLEGCIRTL RYFAGWADKI QGKTIPTDDN VVCFTRHEPI GVCGAITPWN FPLLMLVWKL APALCCGNTM VLKPAEQTPL TALYLGSLIK EAGFPPGVVN IVPGFGPTVG AAISSHPQIN KIAFTGSTEV GKLVKEAASR SNLKRVTLEL GGKNPCIVCA DADLDLAVEC AHQGVFFNQG QCCTAASRVF VEEQVYSEFV RRSVEYAKKR PVGDPFDVKT EQGPQIDQKQ FDKILELIES GKKEGAKLEC GGSAMEDKGL FIKPTVFSEV TDNMRIAKEE IFGPVQPILK FKSIEEVIKR ANSTDYGLTA AVFTKNLDKA LKLASALESG TVWINCYNAL YAQAPFGGFK MSGNGRELGE YALAEYTEVK TVTIKLGDKN P
Greater than 90% as determined by SDS-PAGE.
220
200 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers