missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human AKR1A1 Protein

Product Code. 18177369 Shop All Bio Techne Products
Change view
Click to view available options
:
0.1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
18177369 0.1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18177369 Supplier Novus Biologicals™ Supplier No. NBP130170

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly purified. Generating reliable and reproducible results.

An un-tagged recombinant protein corresponding to the amino acids 1-325 of Human AKR1A1 The Recombinant Human AKR1A1 Protein is derived from E. coli. The Recombinant Human AKR1A1 Protein has been validated for the following applications: SDS-Page.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P14550
For Use With (Application) ELISA, SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH8.0), 50mM NaCl, 10% glycerol
Gene ID (Entrez) 10327
Molecular Weight (g/mol) 36.5kDa
Name AKR1A1 Protein
Purification Method Protein
Quantity 0.1mg
Source E. Coli
Immunogen AKR1A1, 1-325aa, Human
Storage Requirements −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >90%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.