missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ADP-ribosylation factor 4 Protein
A DNA sequence encoding the Homo sapiens (Human) ADP-ribosylation factor 4, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8338-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001651.1 | |
ADP-ribosylation factor 4 Protein | |
Research Use Only | |
ARF2 | |
Recombinant Protein | |
E. coli | |
MGLTISSLFS RLFGKKQMRI LMVGLDAAGK TTILYKLKLG EIVTTIPTIG FNVETVEYKN ICFTVWDVGG QDRIRPLWKH YFQNTQGLIF VVDSNDRERI QEVADELQKM LLVDELRDAV LLLFANKQDL PNAMAISEMT DKLGLQSLRN RTWYVQATCA TQGTGLYEGL DWLSNELSKR | |
Greater than 90% as determined by SDS-PAGE. |
378 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |