missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Actin, cytoplasmic 1 Protein

A DNA sequence encoding the Homo sapiens (Human) Actin, cytoplasmic 1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8379-ec-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16014311

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_001092.1
Actin, cytoplasmic 1 Protein
Research Use Only
ACTB
Recombinant Protein
E. coli
PVLLTEAPLN PKANREKMTQ IMFETFNTPA MYVAIQAVLS LYASGRTTGI VMDSGDGVTH TVPIYEGYAL PHAILRLDLA GRDLTDYLMK ILTERGYSFT TTAEREIVRD IKEKLCYVAL DFEQEMATAA SSSSLEKSYE LPDGQVITIG NERFRCPEAL FQPSFLGMES CGIHETTFNS IMKCDVDIRK DLYANTVLSG GTTMYPGIAD RMQKEITALA PSTMKIKIIA PPERKYSVWI GGSILASLST FQQMWISKQE YDESGPSIVH RKCF
Greater than 90% as determined by SDS-PAGE.
60
10 μg
Not determined.
Partial (see sequence information for more details).
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers