missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ACSS1, mitochondrial Protein
A DNA sequence encoding the Homo sapiens (Human) ACSS1, mitochondrial, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9911-ye-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001239604.1 | |
ACSS1, mitochondrial Protein | |
Research Use Only | |
ACSS1 | |
Recombinant Protein | |
Yeast | |
ASGPSGSAPA VAAAAAQPGS YPALSAQAAR EPAAFWGPLA RDTLVWDTPY HTVWDCDFST GKIGWFLGGQ LNVSVNCLDQ HVRKSPESVA LIWERDEPGT EVRITYRELL ETTCRLANTL KRHGVHRGDR VAIYMPVSPL AVAAMLACAR IGAVHTVIFA GFSAESLAGR INDAKCKVVI TFNQGLRGGR VVELKKIVDE AVKHCPTVQH VLVAHRTDNK VHMGDLDVPL EQEMAKEDPV CAPESMGSED MLFMLYTSGS TGMPKGIVHT QAGYLLYAAL THKLVFDHQP GDIFGCVADI GWITGHSYVV YGPLCNGATS VLFESTPVYP NAGRYWETVE RLKINQFYGA PTAVRLLLKY GDAWVKKYDR SSLRTLGSVG EPINCEAWEW LHRVVGDSRC TLVDTWWQTE TGGICIAPRP SEEGAEILPA MAMRPFFGIV PVLMDEKGSV VEGSNVSGAL CISQAWPGMA RTIYGDHQRF VDAYFKAYPG YYFTGDGAYR TEGGYYQITG RMDDVINISG HRLGTAEIED AIADHPAVPE SAVIGYPHDI KGEAAFAFIV VKDSAGDSDV VVQELKSMVA TKIAKYAVPD EILVVKRLPK TRSGKVMRRL LRKIITSEAQ ELGDTTTLED PSIIAEILSV YQKCKDKQAA AK | |
Greater than 94.5%% as determined by SDS-PAGE. |
84532 | |
50 μg | |
Not determined. | |
Full. Length. of. Mature. Protein. | |
Human | |
His | |
Tris-based buffer, 50% glycerol |