missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human 60S ribosomal protein L35 Protein

A DNA sequence encoding the Homo sapiens (Human) 60S ribosomal protein L35, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8470-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16004791

  • 5325.00 NOK / 100µg
Estimated Shipment: 10-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_009140.1
60S ribosomal protein L35 Protein
Research Use Only
RPL35
Recombinant Protein
E. coli
AKIKARDLRG KKKEELLKQL DDLKVELSQL RVAKVTGGAA SKLSKIRVVR KSIARVLTVI NQTQKENLRK FYKGKKYKPL DLRPKKTRAM RRRLNKHEEN LKTKKQQRKE RLYPLRKYAV KA
Greater than 90% as determined by SDS-PAGE.
11224
100 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers