missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human 60S ribosomal protein L3 Protein
A DNA sequence encoding the Homo sapiens (Human) 60S ribosomal protein L3, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8406-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_000958.1 | |
60S ribosomal protein L3 Protein | |
Research Use Only | |
RPL3 | |
Recombinant Protein | |
E. coli | |
SHRKFSAPRH GSLGFLPRKR SSRHRGKVKS FPKDDPSKPV HLTAFLGYKA GMTHIVREVD RPGSKVNKKE VVEAVTIVET PPMVVVGIVG YVETPRGLRT FKTVFAEHIS DECKRRFYKN WHKSKKKAFT KYCKKWQDED GKKQLEKDFS SMKKYCQVIR VIAHTQMRLL PLRQKKAHLM EIQVNGGTVA EKLDWARERL EQQVPVNQVF GQDEMIDVIG VTKGKGYKGV TSRWHTKKLP RKTHRG | |
Greater than 90% as determined by SDS-PAGE. |
6122 | |
10 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |