missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human 60S ribosomal protein L28 Protein
A DNA sequence encoding the Homo sapiens (Human) 60S ribosomal protein L28, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6619-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_000982.2 | |
60S ribosomal protein L28 Protein | |
Research Use Only | |
RPL28 | |
Recombinant Protein | |
E. coli | |
SAHLQWMVVR NCSSFLIKRN KQTYSTEPNN LKARNSFRYN GLIHRKTVGV EPAADGKGVV VVIKRRSGQR KPATSYVRTT INKNARATLS SIRHMIRKNK YRPDLRMAAI RRASAILRSQ KPVMVKRKRT RPTKSS | |
Greater than 90% as determined by SDS-PAGE. |
6158 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |