missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human 60S acidic ribosomal protein P0 Protein
A DNA sequence encoding the Homo sapiens (Human) 60S acidic ribosomal protein P0, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6623-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_000993.1 | |
60S acidic ribosomal protein P0 Protein | |
Research Use Only | |
RPLP0 | |
Recombinant Protein | |
E. coli | |
MPREDRATWK SNYFLKIIQL LDDYPKCFIV GADNVGSKQM QQIRMSLRGK AVVLMGKNTM MRKAIRGHLE NNPALEKLLP HIRGNVGFVF TKEDLTEIRD MLLANKVPAA ARAGAIAPCE VTVPAQNTGL GPEKTSFFQA LGITTKISRG TIEILSDVQL IKTGDKVGAS EATLLNMLNI SPFSFGLVIQ QVFDNGSIYN PEVLDITEET LHSRFLEGVR NVASVCLQIG YPTVASVPHS IINGYKRVLA LSVETDYTFP LAEKVKAFLA DPSAFVAAAP VAAATTAAPA AAAAPAKVEA KEESEESDED MGFGLFD | |
Greater than 90% as determined by SDS-PAGE. |
6175 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |