missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human 40S ribosomal protein S13 Protein
A DNA sequence encoding the Homo sapiens (Human) 40S ribosomal protein S13, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8411-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001008.1 | |
40S ribosomal protein S13 Protein | |
Research Use Only | |
RPS13 | |
Recombinant Protein | |
E. coli | |
RMHAPGKGLS QSALPYRRSV PTWLKLTSDD VKEQIYKLAK KGLTPSQIGV ILRDSHGVAQ VRFVTGNKIL RILKSKGLAP DLPEDLYHLI KKAVAVRKHL ERNRKDKDAK FRLILIESRI HRLARYYKTK RVLPPNWKYE SSTASALVA | |
Greater than 80% as determined by SDS-PAGE. |
6207 | |
50 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |