missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human 40S ribosomal protein S11 Protein
A DNA sequence encoding the Homo sapiens (Human) 40S ribosomal protein S11, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8389-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001006.1 | |
40S ribosomal protein S11 Protein | |
Research Use Only | |
RPS11 | |
Recombinant Protein | |
E. coli | |
ADIQTERAYQ KQPTIFQNKK RVLLGETGKE KLPRYYKNIG LGFKTPKEAI EGTYIDKKCP FTGNVSIRGR ILSGVVTKMK MQRTIVIRRD YLHYIRKYNR FEKRHKNMSV HLSPCFRDVQ IGDIVTVGEC RPLSKTVRFN VLKVTKAAGT KKQFQKF | |
Greater than 90% as determined by SDS-PAGE. |
6205 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |