missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Hepatitis C Virus polyProtein
A DNA sequence encoding the Hepatitis C virus genotype 1a (isolate 1) (HCV) Hepatitis C Virus polyprotein, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8939-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
Hepatitis C Virus polyProtein | |
Research Use Only | |
Partial (see sequence information for more details). | |
Hepatitis | |
His | |
Tris-based buffer, 50% glycerol |
200 μg | |
Not determined. | |
Recombinant Protein | |
E. coli | |
APITAYAQQT RGLLSTIVTS LTGRDTNENC GEVQVLSTAT QSFLGTAVNG VMWTVYHGAG SKTISGPQGP VNQMYTNVDQ DLVGWPAPPG VRSLAPCTCG ASDLYLVTRH ADVVPVRRRG DTRGALLSPR PISTLKGSSG GPLLCPMGHA AGIFRAAVCT RGVAKAVDFV PVESLETTMR SPVFTDNSTP PAVPQTYQVA HLHAPTGSGK STKVPAAYAA QGYKVLVLNP SVAATLGFGA YMSKAYGIDP NIRTGVRTIT TGAPITYSTY GKFLADGGCS GGAYDIIICD ECHSTDATTI LGIGTALDQA ETAGMRLVVL ATATPPGSVT TPHSNIEEVA LPTTGEVPFY GRAIPLGLIK GGRHLIFCHS KKKCDELAGQ LTSLGLNAVA YYRGLDVSVI PTSGDVVVCA TDALMTGFTG DFDSVIDCNT SVIQTIDFSL DPTFSIETTT VPQDAVSRSQ RRGRTGRGRL GIYRYVTPGE RPSGIFDTAV LCECYDAGCA WYELTPAETT VRLRAYFNTP GLPVCQDHLE FWESVFTGLT NIDGHFLSQT KQSGENFPYL VAYQATVCAR ALAPPPSWDT IWKCLTRLKP TLHGPTPLLY RLGSVQNEVT LTHPITKYIM ACMSADLEVV T | |
Greater than 90% as determined by SDS-PAGE. |