missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Ebola Virus Ebola Sudan Protein
A cDNA sequence encoding the Ebola Sudan Protein was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11726-100ug
Additional Details : Weight : 0.01000kg
Specifications
Ebola Sudan Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
Ebola Sudan protein solution is supplied in Phosphate buffer and 0.02% sodium azide. |
100 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Ebola Virus | |
Untagged | |
>95% pure as determined by 12% SDS-PAGE (Coomassie blue stain). |