missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli Phosphoserine phosphatase Protein
A DNA sequence encoding the Escherichia coli (strain K12) Phosphoserine phosphatase, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6548-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_418805.1 | |
Phosphoserine phosphatase Protein | |
Research Use Only | |
serB | |
Recombinant Protein | |
E. coli | |
MPNITWCDLP EDVSLWPGLP LSLSGDEVMP LDYHAGRSGW LLYGRGLDKQ RLTQYQSKLG AAMVIVAAWC VEDYQVIRLA GSLTARATRL AHEAQLDVAP LGKIPHLRTP GLLVMDMDST AIQIECIDEI AKLAGTGEMV AEVTERAMRG ELDFTASLRS RVATLKGADA NILQQVRENL PLMPGLTQLV LKLETLGWKV AIASGGFTFF AEYLRDKLRL TAVVANELEI MDGKFTGNVI GDIVDAQYKA KTLTRLAQEY EIPLAQTVAI GDGANDLPMI KAAGLGIAYH AKPKVNEKAE VTIRHADLMG VFCILSGSLN QK | |
Greater than 90% as determined by SDS-PAGE. |
948913 | |
500 μg | |
Not determined. | |
Full Length | |
E. coli | |
His-SUMO | |
Tris-based buffer, 50% glycerol |