Learn More
enQuireBio™ Recombinant E. coli Oxygen-insensitive NADPH nitroreductase Protein
A DNA sequence encoding the Escherichia coli (strain K12) Oxygen-insensitive NADPH nitroreductase, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7087-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_415372.1 | |
Oxygen-insensitive NADPH nitroreductase Protein | |
Research Use Only | |
nfsA | |
Recombinant Protein | |
E. coli | |
MTPTIELICG HRSIRHFTDE PISEAQREAI INSARATSSS SFLQCSSIIR ITDKALREEL VTLTGGQKHV AQAAEFWVFC ADFNRHLQIC PDAQLGLAEQ LLLGVVDTAM MAQNALIAAE SLGLGGVYIG GLRNNIEAVT KLLKLPQHVL PLFGLCLGWP ADNPDLKPRL PASILVHENS YQPLDKGALA QYDEQLAEYY LTRGSNNRRD TWSDHIRRTI IKESRPFILD YLHKQGWATR | |
Greater than 90% as determined by SDS-PAGE. |
945483 | |
500 μg | |
Not determined. | |
Full Length | |
E. coli | |
His-SUMO | |
Tris-based buffer, 50% glycerol |