missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant E. coli Outer-membrane lipoprotein carrier Protein

A DNA sequence encoding the Escherichia coli O9:H4 (strain HS) Outer-membrane lipoprotein carrier protein, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7432-ec-200ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16018850

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

WP_001295343.1
200 μg
Not determined.
Full Length
E. coli
His
Tris-based buffer, 50% glycerol
Outer-membrane lipoprotein carrier Protein
Research Use Only
lolA
Recombinant Protein
E. coli
DAASDLKSRL DKVSSFHASF TQKVTDGSGA AVQEGQGDLW VKRPNLFNWH MTQPDESILV SDGKTLWFYN PFVEQATATW LKDATGNTPF MLIARNQSSD WQQYNIKQNG DDFVLTPKAS NGNLKQFTIN VGRDGTIHQF SAVEQDDQRS SYQLKSQQNG AVDAAKFTFT PPQGVTVDDQ RK
Greater than 90% as determined by SDS-PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers