missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli L-lactate dehydrogenase Protein
A DNA sequence encoding the Escherichia coli O9:H4 (strain HS) L-lactate dehydrogenase, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7434-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
WP_000586962.1 | |
200 μg | |
Not determined. | |
Full Length | |
E. coli | |
His | |
Tris-based buffer, 50% glycerol |
L-lactate dehydrogenase Protein | |
Research Use Only | |
lldD | |
Recombinant Protein | |
E. coli | |
MIISAASDYR AAAQRILPPF LFHYMDGGAY SEYTLRRNVE DLSEVALRQR ILKNMSDLSL ETTLFNEKLS MPVALAPVGL CGMYARRGEV QAAKAADAHG IPFTLSTVSV CPIEEVAPAI KRPMWFQLYV LRDRGFMRNA LERAKAAGCS TLVFTVDMPT PGARYRDAHS GMSGPNAAMR RYLQAVTHPQ WAWDVGLNGR PHDLGNISAY LGKPTGLEDY IGWLGNNFDP SISWKDLEWI RDFWDGPMVI KGILDPEDAR DAVRFGADGI VVSNHGGRQL DGVLSSARAL PAIADAVKGD IAILADSGIR NGLDVVRMIA LGADTVLLGR AFLYALATAG QAGVANLLNL IEKEMKVAMT LTGAKSISEI TQDSLVQGLG KELPAALAPM AKGNAA | |
Greater than 90% as determined by SDS-PAGE. |