missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli Elongation factor P Protein
A DNA sequence encoding the Escherichia coli (strain K12) Elongation factor P, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7316-ye-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_418571.1 | |
Elongation factor P Protein | |
Research Use Only | |
efp | |
Recombinant Protein | |
Yeast | |
ATYYSNDFRA GLKIMLDGEP YAVEASEFVK PGKGQAFARV KLRRLLTGTR VEKTFKSTDS AEGADVVDMN LTYLYNDGEF WHFMNNETFE QLSADAKAIG DNAKWLLDQA ECIVTLWNGQ PISVTPPNFV ELEIVDTDPG LKGDTAGTGG KPATLSTGAV VKVPLFVQIG EVIKVDTRSG EYVSRVK | |
Greater than 90% as determined by SDS-PAGE. |
948661 | |
500 μg | |
Not determined. | |
Full Length | |
E. coli | |
His | |
Tris-based buffer, 50% glycerol |