missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant E. coli CFA / I fimbrial subunit E Protein

A DNA sequence encoding the Escherichia coli CFA/I fimbrial subunit E, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7131-ec-200ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16017470

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

WP_001033658.1
200 μg
Not determined.
Full Length
E. coli
His-SUMO
Tris-based buffer, 50% glycerol
CFA / I fimbrial subunit E Protein
Research Use Only
cfaE
Recombinant Protein
E. coli
MNKILFIFTL FFSSGFFTFA VSADKNPGSE NMTNTIGPHD RGGSSPIYNI LNSYLTAYNG SHHLYDRMSF LCLSSQNTLN GACPSSDAPG TATIDGETNI TLQFTEKRSL IKRELQIKGY KQFLFKNANC PSKLALNSSH FQCNREQASG ATLSLYIPAG ELNKLPFGGV WNAVLKLNVK RRYDTTYGTY TINITVNLTD KGNIQIWLPQ FKSNARVDLN LRPTGGGTYI GRNSVDMCFY DGYSTNSSSL EIRFQDDNSK SDGKFYLKKI NDDSKELVYT LSLLLAGKNL TPTNGQALNI NTASLETNWN RITAVTMPEI SVPVLCWPGR LQLDAKVKNP EAGQYMGNIK ITFTPSSQTL
Greater than 80% as determined by SDS-PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers