missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli Cell division protein FtsZ Protein
A DNA sequence encoding the Escherichia coli O6:H1 (strain CFT073 / ATCC 700928 / UPEC) Cell division protein FtsZ, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7325-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
WP_000462776.1 | |
50 μg | |
Not determined. | |
Full Length | |
E. coli | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
Cell division protein FtsZ Protein | |
Research Use Only | |
ftsZ | |
Recombinant Protein | |
E. coli | |
MFEPMELTND AVIKVIGVGG GGGNAVEHMV RERIEGVEFF AVNTDAQALR KTAVGQTIQI GSGITKGLGA GANPEVGRNA ADEDRDALRA ALEGADMVFI AAGMGGGTGT GAAPVVAEVA KDLGILTVAV VTKPFNFEGK KRMAFAEQGI TELSKHVDSL ITIPNDKLLK VLGRGISLLD AFGAANDVLK GAVQGIAELI TRPGLMNVDF ADVRTVMSEM GYAMMGSGVA SGEDRAEEAA EMAISSPLLE DIDLSGARGV LVNITAGFDL RLDEFETVGN TIRAFASDNA TVVIGTSLDP DMNDELRVTV VATGIGMDKR PEITLVTNKQ VQQPVMDRYQ QHGMAPLTQE QKPVAKVVND NAPQTAKEPD YLDIPAFLRK QAD | |
Greater than 90% as determined by SDS-PAGE. |