missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli ATP-dependent DNA helicase Rep Protein
A DNA sequence encoding the Escherichia coli (strain K12) ATP-dependent DNA helicase Rep, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8666-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
WP_001238899.1 | |
ATP-dependent DNA helicase Rep Protein | |
Research Use Only | |
rep | |
Recombinant Protein | |
E. coli | |
MRLNPGQQQA VEFVTGPCLV LAGAGSGKTR VITNKIAHLI RGCGYQARHI AAVTFTNKAA REMKERVGQT LGRKEARGLM ISTFHTLGLD IIKREYAALG MKANFSLFDD TDQLALLKEL TEGLIEDDKV LLQQLISTIS NWKNDLKTPS QAAASAIGER DRIFAHCYGL YDAHLKACNV LDFDDLILLP TLLLQRNEEV RKRWQNKIRY LLVDEYQDTN TSQYELVKLL VGSRARFTVV GDDDQSIYSW RGARPQNLVL LSQDFPALKV IKLEQNYRSS GRILKAANIL IANNPHVFEK RLFSELGYGA ELKVLSANNE EHEAERVTGE LIAHHFVNKT QYKDYAILYR GNHQSRVFEK FLMQNRIPYK ISGGTSFFSR PEIKDLLAYL RVLTNPDDDS AFLRIVNTPK REIGPATLKK LGEWAMTRNK SMFTASFDMG LSQTLSGRGY EALTRFTHWL AEIQRLAERE PIAAVRDLIH GMDYESWLYE TSPSPKAAEM RMKNVNQLFS WMTEMLEGSE LDEPMTLTQV VTRFTLRDMM ERGESEEELD QVQLMTLHAS KGLEFPYVYM VGMEEGFLPH QSSIDEDNID EERRLAYVGI TRAQKELTFT LCKERRQYGE LVRPEPSRFL LELPQDDLIW EQERKVVSAE ERMQKGQSHL ANLKAMMAAK RGK | |
Greater than 90% as determined by SDS-PAGE. |
948292 | |
50 μg | |
Not determined. | |
Full Length | |
E. coli | |
GST | |
Tris-based buffer, 50% glycerol |