Learn More
enQuireBio™ Recombinant E. coli 50S ribosomal protein L6 Protein
A DNA sequence encoding the Escherichia coli (strain K12) 50S ribosomal protein L6, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8624-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_417764.1 | |
50S ribosomal protein L6 Protein | |
Research Use Only | |
rplF | |
Recombinant Protein | |
E. coli | |
SRVAKAPVVV PAGVDVKING QVITIKGKNG ELTRTLNDAV EVKHADNTLT FGPRDGYADG WAQAGTARAL LNSMVIGVTE GFTKKLQLVG VGYRAAVKGN VINLSLGFSH PVDHQLPAGI TAECPTQTEI VLKGADKQVI GQVAADLRAY RRPEPYKGKG VRYADEVVRT KEAK | |
Greater than 90% as determined by SDS-PAGE. |
947803 | |
500 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
E. coli | |
GST | |
Tris-based buffer, 50% glycerol |