missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant E. coli 30S ribosomal protein S15 Protein

A DNA sequence encoding the Escherichia coli (strain K12) 30S ribosomal protein S15, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8619-ec-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16045611

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_417634.1
30S ribosomal protein S15 Protein
Research Use Only
rpsO
Recombinant Protein
E. coli
SLSTEATAKI VSEFGRDAND TGSTEVQVAL LTAQINHLQG HFAEHKKDHH SRRGLLRMVS QRRKLLDYLK RKDVARYTQL IERLGLRR
Greater than 90% as determined by SDS-PAGE.
947686
50 μg
Not determined.
Full Length
E. coli
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers