missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Dog LH-beta Protein
A DNA sequence encoding the Canis lupus familiaris (Dog) (Canis familiaris) LH-beta, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6304-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001183962.1 | |
LH-beta Protein | |
Research Use Only | |
LHB | |
Recombinant Protein | |
E. coli | |
SRGPLRPLCR PINATLAAEN EACPVCITFT TTICAGYCPS MVRVLPAALP PVPQPVCTYH ELHFASIRLP GCPPGVDPMV SFPVALSCRC GPCRLSNSDC GGPRAQSLAC DRPLLPGLLF L | |
Greater than 90% as determined by SDS-PAGE. |
403959 | |
50 μg | |
Not determined. | |
Full Length of Mature Protein | |
Dog | |
His | |
Tris-based buffer, 50% glycerol |