missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Dog Annexin A4 Protein
A DNA sequence encoding the Canis lupus familiaris (Dog) (Canis familiaris) Annexin A4, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5653-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
Annexin A4 Protein | |
Research Use Only | |
ANXA4 | |
Recombinant Protein | |
E. coli | |
AAKGGTVKPA SGFSATEDAQ TLRKAMKGLG TDEDAIISVL APRNTSQRQE IRTAYKSTIG RDLMDDLKSE LSGNFERVIV GMITPTVLYD VQELRRAMKG SGTDEGCLIE ILASRTPEEL RCINQTYQLQ YGRSLEDVIR SDTSFMFQRV LVSLSAGGRD EGNFLDDALM RQDAQDLYEA GEKKWGTDEV KFLTVLCSRN RNHLLHVFDE YKRISQKDIE QGIKSETSGS FEDALLAIVK CMRNKSAYFA ERLYKSMKGL GTDDNTLIRV MVSRAEIDMM DIRESFKRLY GKSLYSFIKG DTSGDYRKVL LILCGGDD | |
Greater than 90% as determined by SDS-PAGE. |
100 μg | |
Not determined. | |
Full Length | |
Dog | |
His-SUMO | |
Tris-based buffer, 50% glycerol |