missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Chinese Hamster Peroxiredoxin 1 / PRDX1 Protein
A DNA sequence encoding the Cricetulus griseus (Chinese hamster) (Cricetulus barabensis griseus) Peroxiredoxin 1 / PRDX1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8117-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001233694.1 | |
Peroxiredoxin 1 / PRDX1 Protein | |
Research Use Only | |
PRDX1 | |
Recombinant Protein | |
E. coli | |
MSSGNAKIGY PAPNFKATAV MPDGQFRDIC LSEYRGKYVV FFFYPLDFTF VCPTEIIAFS DRAEEFKKLN CQVIGASVDS HFCHLAWINT PKKQGGLGPM NIPLVSDPKR TIAQDYGVLK ADEGISFRGL FIIDDKGILR QITINDLPVG RSVDEILRLV QAFQFTDKHG EVCPAGWKPG SDTIKPDVQK SKEYFSKQK | |
Greater than 90% as determined by SDS-PAGE. |
100689332 | |
200 μg | |
Not determined. | |
Full Length | |
Chinese Hamster | |
His | |
Tris-based buffer, 50% glycerol |