missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Chicken TGF-beta 1 / TGFB1 Protein
A DNA sequence encoding the Gallus gallus (Chicken) TGF-beta 1 / TGFB1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6779-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
TGF-beta 1 / TGFB1 Protein | |
Research Use Only | |
TGFB1 | |
Recombinant Protein | |
E. coli | |
DLDTDYCFGP GTDEKNCCVR PLYIDFRKDL QWKWIHEPKG YMANFCMGPC PYIWSADTQY TKVLALYNQH NPGASAAPCC VPQTLDPLPI IYYVGRNVRV EQLSNMVVRA CKCS | |
Greater than 90% as determined by SDS-PAGE. |
500 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Chicken | |
His-SUMO | |
Tris-based buffer, 50% glycerol |