missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Bovine Decorin / DCN / SLRR1B Protein
A DNA sequence encoding the Bos taurus (Bovine) Decorin / DCN / SLRR1B, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9101-ye-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_776331.2 | |
Decorin / DCN / SLRR1B Protein | |
Research Use Only | |
DCN | |
Recombinant Protein | |
Yeast | |
DEASGIGPEE HFPEVPEIEP MGPVCPFRCQ CHLRVVQCSD LGLEKVPKDL PPDTALLDLQ NNKITEIKDG DFKNLKNLHT LILINNKISK ISPGAFAPLV KLERLYLSKN QLKELPEKMP KTLQELRVHE NEITKVRKSV FNGLNQMIVV ELGTNPLKSS GIENGAFQGM KKLSYIRIAD TNITTIPQGL PPSLTELHLD GNKITKVDAA SLKGLNNLAK LGLSFNSISA VDNGSLANTP HLRELHLNNN KLVKVPGGLA DHKYIQVVYL HNNNISAIGS NDFCPPGYNT KKASYSGVSL FSNPVQYWEI QPSTFRCVYV RAAVQLGNYK | |
Greater than 90% as determined by SDS-PAGE. |
280760 | |
50 μg | |
Not determined. | |
Full Length | |
Bovine | |
His | |
Tris-based buffer, 50% glycerol |