Learn More
GFL Gesellschaft fuer Labortec™ Reciprocating Shaker
Product Code. 10100841
Change view
Click to view available options
Speed Range:
20 to 300 min-1
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Speed Range Unit Size
10100841 20 to 300 min-1 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10100841 Supplier GFL Gesellschaft fuer Labortec™ Supplier No. 3016
Quantity Price
1 25604.44 NOK / Pack of 1
3 21474.45 NOK / Pack of 1
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Use these shakers in research labs, standard labs and special labs for medicine, science and industry. GFL Gesellschaft fuer Labortec™ Reciprocating Shakers are ideal for vigorous and intensive shaking of clamped lying flasks, mixing flasks or separating funnels in continuous operation.
  • Compact, low-wear counterbalanced drive mechanism, ensuring dependability for slow rocking motion
  • Housing made of high impact strength polystyrene, off-white varnish
  • Base plate is made of electrolytically galvanized, powder-coated steel sheet, the shaking platform of anodized aluminum, equipped with four plastic pins for secure attachment of accessory equipment
  • A.C. motor with overload protection drives the unit
  • Clearly laid-out control panel for easy operation
  • Electronic speed control, stepless, gentle start-up
  • Constant speed during continuous operation, independent of load
Specifications
Speed Range 20 to 300 min-1
Electrical Requirements 115/230 V, 50/60 Hz, 65 W
Product Type Reciprocating Shaker
Dimensions (LxW) Platform 450 x 450 mm
Dimensions (L x W x H) 625 x 510 x 145 mm
Shipping Weight (Metric) 25 kg
Timer Range Up to 60 min., Continuous