Learn More
UVP Rechargeable UV Lamp
Specifications
Specifications
| Wattage | 6 W |
| Wavelength | 365/254 nm |
| Dimensions (L x W x H) | 249 x 76 x 140 mm |
| Height (Metric) | 140 mm |
| Length (Metric) | 249 mm |
| Model | UVSL-26P |
| Width (Metric) | 76 mm |
| Product Type | Rechargeable UV Lamp |