Learn More
Lenz Laborglasinstrumente™ Receiving Flask with PUR Coating
Product Code. 10355385
Change view
Click to view available options
Volume (Metric):
500 mL
1000 mL
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
10355385 1000 mL Each
12343443 500 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10355385 Supplier Lenz Laborglasinstrumente™ Supplier No. 03064070
  • ONLINE EXCLUSIVE PRICE
    1645.00 NOK
    1215.05 NOK / Pack of 1
    Save 429.95 NOK
    26% Off
Estimated Shipment: 29-12-2026
Especially for use in rotary evaporators. Lenz Laborglasinstrumente™ Receiving Flask with PUR Coating are manufactured from durable DURAN™ glass.
  • With spherical joint S 35
  • With highly transparent PUR coating
  • Autoclavable
  • Available in a range of capacities
Specifications
Material Duran Glass
Type Receiving Flask
For Use With (Equipment) Rotary Evaporators
Material (Coating) PUR
Volume (Metric) 1000 mL
Diameter (Metric) 131 mm