Learn More
Cole-Parmer™ Stuart™ Receiving Flask For Stuart Rotary Evaporator
Product Code. 12497802
Change view
Click to view available options
Volume (Metric):
50 mL
100 mL
Unit Size:
Pack of 2
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
12497802 50 mL Pack of 2
12477802 100 mL Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12497802 Supplier Cole-Parmer™ Supplier No. 1009671
  • ONLINE EXCLUSIVE PRICE
    1120.00 NOK
    996.09 NOK / Pack of 2
    Save 123.91 NOK
    11% Off
 Restricted Item
Estimated Shipment: 24-11-2026
Can be used with all Stuart™ glassware sets
  • For use with Stuart™ Rotary Evaporator
  • S35 spherical socket
  • Round bottom
Specifications
Type Receiving Flask
For Use With (Equipment) Stuart Rotary Evaporators
Shape Spherical
Taper S35 spherical socket
Volume (Metric) 50 mL